Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,4,5-trifluorobenzene, 98+%

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene
CAS 327-52-6
Molecular Formula C6H2BrF3
Formula Weight 210.98
MDL Number MFCD00000306
UN Number UN1993
Beilstein 1636588
Synonym 2,4,5-Trifluorobromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1196203

  SDS
alfa aesar™
A1196203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1196214

  SDS
alfa aesar™
A1196214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.98
Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene
Assay ≥98%
Molecular Formula C6H2BrF3
MDL Number MFCD00000306
CAS 327-52-6
UN Number UN1993
Melting Point −19°C
Beilstein 1636588
Synonym 2,4,5-Trifluorobromobenzene
Boiling Point 143° to 144°C
Quantity 1g
Flash Point 55°C (131°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Applications
1-Bromo-2,4,5-trifluorobenzene is the intermediate of liquid crystals. The derivatives of this compound can be used as intermediates for the synthesis of qunolone antibacterial. It is employed in the synthesis of 3-ethenyl, 3-ethynyl, 3-aryl, and 3-cyclopropyl-2, 4, 5-trifluorobenzoic acids from 1-bromo-2, 4, 5-trifluorobenzene and 2, 4, 5-trifluoro-3-hydroxybenzoic acid.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.