Please login to your online account to display your discounted pricing

1-Bromo-2,3-difluorobenzene, 98%, ACROS Organics™

$54.15 - $54.15

Chemical Identifiers

Chemical Name or Material 1-Bromo-2, 3-difluorobenzene
CAS 38573-88-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3BrF2
Linear Formula BrC6H3F2
Formula Weight 193
MDL Number MFCD00061136
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308670010

  SDS
thermo fisher scientific
308670010
Glass bottle 1g Each for $54.15
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 193
Quantity 1g
Color Clear colorless to peach
CAS Max % 100.0
Chemical Name or Material 1-Bromo-2, 3-difluorobenzene
Linear Formula BrC6H3F2
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00061136
CAS 38573-88-5
Packaging Glass bottle
Refractive Index 1.5090 to 1.5100 (20°C, 589nm)
Molecular Formula C6H3BrF2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.