missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,3,5-trifluorobenzene, TCI America™

$62.64 - $190.97

Chemical Identifiers

CAS 133739-70-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00012232
InChI Key XSMLLZPSNLQCQU-UHFFFAOYSA-N
Synonym 2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
PubChem CID 2736399
IUPAC Name 1-bromo-2,3,5-trifluorobenzene
SMILES C1=C(C=C(C(=C1F)F)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29701G View Documents TCI America
B29701G
1g Each for $62.64
 
B29705G View Documents TCI America
B29705G
5g Each for $190.97
 
Specifications

Chemical Identifiers

133739-70-5
210.981
XSMLLZPSNLQCQU-UHFFFAOYSA-N
2736399
C1=C(C=C(C(=C1F)F)Br)F
C6H2BrF3
MFCD00012232
2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
1-bromo-2,3,5-trifluorobenzene

Specifications

Yellow
133739-70-5
MFCD00012232
2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
C1=C(C=C(C(=C1F)F)Br)F
210.981
210.98
≥98.0% (GC)
1g
C6H2BrF3
1993
XSMLLZPSNLQCQU-UHFFFAOYSA-N
1-bromo-2,3,5-trifluorobenzene
2736399
Clear Liquid at 20°C
1-Bromo-2,3,5-trifluorobenzene
Safety and Handling

Safety and Handling

TSCA : No