missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,3,5-trichlorobenzene, TCI America™

$58.06 - $169.59

Chemical Identifiers

CAS 81067-38-1
Molecular Formula C6H2BrCl3
Molecular Weight (g/mol) 260.336
MDL Number MFCD00070734
InChI Key WOIGJFAVHOCDDW-UHFFFAOYSA-N
Synonym 2,3,5-trichlorobromobenzene, benzene, 1-bromo-2,3,5-trichloro, pubchem3192, acmc-209pkn, woigjfavhocddw-uhfffaoysa, benzene,1-bromo-2,3,5-trichloro, 1-bromo-2,3,5-trichlorobenzene, 1-bromanyl-2,3,5-tris chloranyl benzene
PubChem CID 4382903
IUPAC Name 1-bromo-2,3,5-trichlorobenzene
SMILES C1=C(C=C(C(=C1Cl)Cl)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19911G View Documents TCI America
B19911G
1g Each for $58.06
 
B19915G View Documents TCI America
B19915G
5g Each for $169.59
 
Specifications

Chemical Identifiers

81067-38-1
260.336
WOIGJFAVHOCDDW-UHFFFAOYSA-N
4382903
C1=C(C=C(C(=C1Cl)Cl)Br)Cl
C6H2BrCl3
MFCD00070734
2,3,5-trichlorobromobenzene, benzene, 1-bromo-2,3,5-trichloro, pubchem3192, acmc-209pkn, woigjfavhocddw-uhfffaoysa, benzene,1-bromo-2,3,5-trichloro, 1-bromo-2,3,5-trichlorobenzene, 1-bromanyl-2,3,5-tris chloranyl benzene
1-bromo-2,3,5-trichlorobenzene

Specifications

Undesignated
1g
C6H2BrCl3
2,3,5-trichlorobromobenzene, benzene, 1-bromo-2,3,5-trichloro, pubchem3192, acmc-209pkn, woigjfavhocddw-uhfffaoysa, benzene,1-bromo-2,3,5-trichloro, 1-bromo-2,3,5-trichlorobenzene, 1-bromanyl-2,3,5-tris chloranyl benzene
C1=C(C=C(C(=C1Cl)Cl)Br)Cl
260.336
260.34
≥98.0% (GC)
60°C
81067-38-1
MFCD00070734
WOIGJFAVHOCDDW-UHFFFAOYSA-N
1-bromo-2,3,5-trichlorobenzene
4382903
Crystal-Powder at 20°C
1-Bromo-2,3,5-trichlorobenzene
Safety and Handling

Safety and Handling

TSCA : No