Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,3,5-trichlorobenzene, 98%

$31.09 - $426.38

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,3,5-trichlorobenzene
CAS 81067-38-1
Molecular Formula C6H2BrCl3
Formula Weight 260.34
MDL Number MFCD00070734
Beilstein 5926729
Synonym 2,3,5-Trichlorobromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2238403

  SDS
alfa aesar™
B2238403
1g Each for $31.09

AAB2238406

  SDS
alfa aesar™
B2238406
5g Each for $101.28

AAB2238414

  SDS
alfa aesar™
B2238414
25g Each for $426.38
Description & Specifications

Specifications

Formula Weight 260.34
Chemical Name or Material 1-Bromo-2,3,5-trichlorobenzene
Assay 98%
Molecular Formula C6H2BrCl3
MDL Number MFCD00070734
CAS 81067-38-1
Melting Point 58° to 60°C
Beilstein 5926729
Synonym 2,3,5-Trichlorobromobenzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.