missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,3,4-trifluorobenzene, TCI America™

Manufacturer:  TCI America T16781G

 View more versions of this product

Catalog No. T16781G



Specifications

Specifications

1-Bromo-2,3,4-trifluorobenzene
Yellow
176317-02-5
MFCD00013248
MUUAQFJJUGVBGB-UHFFFAOYSA-N
1-bromo-2,3,4-trifluorobenzene
2733255
Clear Liquid at 20°C
60°C
1g
C6H2BrF3
2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f
C1=CC(=C(C(=C1F)F)F)Br
210.981
210.98
≥98.0% (GC)

Chemical Identifiers

176317-02-5
210.981
MUUAQFJJUGVBGB-UHFFFAOYSA-N
2733255
C1=CC(=C(C(=C1F)F)F)Br
C6H2BrF3
MFCD00013248
2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f
1-bromo-2,3,4-trifluorobenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents