Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,3,4-trifluorobenzene, 99%

$145.70 - $145.70

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,3,4-trifluorobenzene
CAS 176317-02-5
Molecular Formula C6H2BrF3
Formula Weight 210.98
MDL Number MFCD00013248
Beilstein 7805451
Synonym 2,3,4-Trifluorobromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1851406

  SDS
alfa aesar™
A1851406
5g Each for $145.70
Description & Specifications

Specifications

Formula Weight 210.98
Synonym 2,3,4-Trifluorobromobenzene
Chemical Name or Material 1-Bromo-2,3,4-trifluorobenzene
Quantity 5g
Health Hazard 1 Warning
Beilstein 7805451
MDL Number MFCD00013248
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 47° to 49°C (60mmHg)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 176317-02-5
Assay 99%
Molecular Formula C6H2BrF3
Flash Point 62°C (143°F)

Applications
1-Bromo-2,3,4-trifluorobenzene is used in synthesis of 5-nitro-6-methylquinoline carboxylic acid.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.