missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,2-dimethoxypropane, 97%, ACROS Organics™

Chemical Identifiers

CAS 126-38-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H11BrO2
Linear Formula CH3C(OCH3)2CH2Br
Formula Weight 183.04
MDL Number MFCD00000208
Synonym Bromoacetone dimethyl ketal
Chemical Name or Material 1-Bromo-2, 2-dimethoxypropane, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC197400100 SDS Thermo Fisher Scientific
197400100
Glass bottle 10mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

126-38-5
100.0
C5H11BrO2
183.04
Bromoacetone dimethyl ketal
≥96.0%
96.0
97%
CH3C(OCH3)2CH2Br
MFCD00000208
1-Bromo-2, 2-dimethoxypropane, 97%

Specifications

126-38-5
100.0
Glass bottle
Liquid
C5H11BrO2
183.04
Clear light yellow
10mL
≥96.0%
96.0
Authentic
1.4465 to 1.4485 (20°C, 589nm)
97%
CH3C(OCH3)2CH2Br
MFCD00000208
Bromoacetone dimethyl ketal
1-Bromo-2, 2-dimethoxypropane, 97%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-784-7

TSCA : TSCA