Please login to your online account to display your discounted pricing

1-Bromo-1-propene, 95%, mixture of isomers, stabilized, ACROS Organics™


Quantity 100mL

Manufacturer: thermo fisher scientific  AC346561000

 View more versions of this product

Catalog No. AC346561000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 120.97
Quantity 100mL
Color Clear colorless to yellow
Density 1.4130g/mL
Beilstein 01, 200
Fieser 13,219
UN Number 1993
CAS Min % 94.0
CAS Max % 100.0
Linear Formula CH3CH=CHBr
MDL Number MFCD00000186
Infrared Spectrum Authentic
Assay Percent Range 95%
CAS 590-14-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Chemical Name or Material 1-Bromo-1-propene
Stabilizer copper
Name Note Mixture of isomers, Stabilized
Molecular Formula C3H5Br
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4520 to 1.4550 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromo-1-propene
Name Note Mixture of isomers, Stabilized
CAS 590-14-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H5Br
Linear Formula CH3CH=CHBr
Formula Weight 120.97
MDL Number MFCD00000186
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents