missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-1-propene, 95%, mixture of isomers, stabilized, ACROS Organics™

Chemical Identifiers

CAS 590-14-7
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C3H5Br
Linear Formula CH3CH=CHBr
Formula Weight 120.97
MDL Number MFCD00000186
Chemical Name or Material 1-Bromo-1-propene, Mixture of isomers, Stabilized
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC346561000 Thermo Fisher Scientific
AC346561000
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

590-14-7
100.0
C3H5Br
120.97
1-Bromo-1-propene, Mixture of isomers, Stabilized
94.0
95%
CH3CH=CHBr
MFCD00000186
≥94%

Specifications

590-14-7
Authentic
1.4520 to 1.4550 (20°C, 589nm)
1.4130g/mL
Liquid
95%
CH3CH=CHBr
MFCD00000186
01, 200
1-Bromo-1-propene, Mixture of isomers, Stabilized
94.0
100.0
copper
100mL
Clear colorless to yellow
C3H5Br
120.97
1993
13,219
≥94%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger