Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-1-chloro-2,2,2-trifluoroethane, 99%

$54.65 - $135.46

Chemical Identifiers

Chemical Name or Material 1-Bromo-1-chloro-2,2,2-trifluoroethane
CAS 151-67-7
Molecular Formula CF3CHBrCl
Formula Weight 197.39
MDL Number MFCD00009602
Beilstein 1736947
Merck Index 14,4603
Synonym 2-Bromo-2-chloro-1,1,1-trifluoroethane; Halothane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1017914

 
alfa aesar™
L1017914
25g Each for $54.65

AAL1017922

 
alfa aesar™
L1017922
100g Each for $135.46
Description & Specifications

Specifications

Formula Weight 197.39
Chemical Name or Material 1-Bromo-1-chloro-2,2,2-trifluoroethane
MDL Number MFCD00009602
CAS 151-67-7
Beilstein 1736947
Synonym 2-Bromo-2-chloro-1,1,1-trifluoroethane; Halothane
Boiling Point 49° to 50°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H341
Causes serious eye damage.
Suspected of causing genetic defects.
Recommended Storage 2° to 8°C
Assay 99%
Molecular Formula CF3CHBrCl
Merck Index 14,4603
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number KH6550000

Applications
1-Bromo-1-chloro-2,2,2-trifluoroethane has been used in a study that combined molecular simulations with free energy simulations to study solvation of halothane in polarizable water and methanol. It is also used in a study to investigate the interaction of anesthetics with the Rho GTPase regulator Rho GDP dissociation inhibitor. In an effect believed to be independent of anesthesia, halothane inhibits synthesis of 5-hydroxytryptamine in brain tissue, probably at the tryptophan hydroxylase step.

Solubility
Soluble in water (8 mg/ml).

Notes
Light Sensitive. Store in the dark. Store away from oxidizing agents. Incompatible with light and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H341
Causes serious eye damage.
Suspected of causing genetic defects.

2° to 8°C