Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-pyrrole-2-carboxaldehyde, 98%

Chemical Identifiers

Chemical Name or Material 1-Boc-pyrrole-2-carboxaldehyde
CAS 161282-57-1
Molecular Formula C10H13NO3
Formula Weight 195.22
MDL Number MFCD03086220
Synonym 1-(tert-Butoxycarbonyl)pyrrole-2-carboxaldehyde; tert-Butyl 2-formylpyrrole-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3255306

  SDS
alfa aesar™
H3255306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.22
MDL Number MFCD03086220
Molecular Formula C10H13NO3
CAS 161282-57-1
Quantity 5g
Chemical Name or Material 1-Boc-pyrrole-2-carboxaldehyde
Synonym 1-(tert-Butoxycarbonyl)pyrrole-2-carboxaldehyde; tert-Butyl 2-formylpyrrole-1-carboxylate
Melting Point 49° to 52°C
Assay 98%
Flash Point 107.0°C (225°F)