Learn More
1-Boc-pyrrole-2-carbonitrile, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1380500-94-6 |
---|---|
Molecular Formula | C10H12N2O2 |
Molecular Weight (g/mol) | 192.218 |
InChI Key | GDHNTBXGTBFNIC-UHFFFAOYSA-N |
Synonym | tert-butyl 2-cyano-1h-pyrrole-1-carboxylate, 1-n-boc-2-cyanopyrrole |
PubChem CID | 86277481 |
IUPAC Name | tert-butyl 2-cyanopyrrole-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1C=CC=C1C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66520MD
|
Thermo Scientific Chemicals
H66520MD |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AAH6652003
|
Thermo Scientific Chemicals
H6652003 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1380500-94-6 | |
192.218 | |
tert-butyl 2-cyano-1h-pyrrole-1-carboxylate, 1-n-boc-2-cyanopyrrole | |
tert-butyl 2-cyanopyrrole-1-carboxylate |
C10H12N2O2 | |
GDHNTBXGTBFNIC-UHFFFAOYSA-N | |
86277481 | |
CC(C)(C)OC(=O)N1C=CC=C1C#N |
Specifications
1380500-94-6 | |
250mg | |
tert-butyl 2-cyano-1h-pyrrole-1-carboxylate, 1-n-boc-2-cyanopyrrole | |
CC(C)(C)OC(=O)N1C=CC=C1C#N | |
192.218 | |
192.21 | |
1-Boc-pyrrole-2-carbonitrile |
C10H12N2O2 | |
UN3439 | |
GDHNTBXGTBFNIC-UHFFFAOYSA-N | |
tert-butyl 2-cyanopyrrole-1-carboxylate | |
86277481 | |
97% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures