missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-pyrrole-2-boronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H64171MD

 View more versions of this product

Catalog No. AAH64171MD



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

1-Boc-pyrrole-2-boronic acid pinacol ester
1072944-98-9
MFCD11504960
DZQURIUJAGTAFW-UHFFFAOYSA-N
tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
46739108
Solid
250mg
C15H24BNO4
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C
293.17
293.17
98%

Chemical Identifiers

1072944-98-9
293.17
DZQURIUJAGTAFW-UHFFFAOYSA-N
46739108
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C
C15H24BNO4
MFCD11504960
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate
tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents