missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-pyrrole-2-boronic acid, 96%

Manufacturer:  Alfa Aesar™ H5307803

 View more versions of this product

Catalog No. AAH5307803



Specifications

Specifications

1-Boc-pyrrole-2-boronic acid
1g
C9H14BNO4
n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid, 1-boc-pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, boc-pyrrole-2-boronic acid, 1-boc-2-pyrrolylboronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid
B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O
211.024
211.02
105°C to 108°C
135884-31-0
MFCD01318939
ZWGMJLNXIVRFRJ-UHFFFAOYSA-N
[1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid
2734321
96%

Chemical Identifiers

135884-31-0
211.024
ZWGMJLNXIVRFRJ-UHFFFAOYSA-N
2734321
B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O
C9H14BNO4
MFCD01318939
n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid, 1-boc-pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, boc-pyrrole-2-boronic acid, 1-boc-2-pyrrolylboronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid
[1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C

Documents