Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-piperidine, 98%

$41.55 - $130.22

Chemical Identifiers

Chemical Name or Material 1-Boc-piperidine
CAS 75844-69-8
Molecular Formula C10H19NO2
Formula Weight 185.27
MDL Number MFCD02093935
Beilstein 3604898
Synonym 1-(tert-Butoxycarbonyl)piperidine; tert-Butyl piperidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1747603

  SDS
alfa aesar™
L1747603
1g Each for $41.55

AAL1747606

  SDS
alfa aesar™
L1747606
5g Each for $130.22
Description & Specifications

Specifications

Formula Weight 185.27
Chemical Name or Material 1-Boc-piperidine
CAS 75844-69-8
Synonym 1-(tert-Butoxycarbonyl)piperidine; tert-Butyl piperidine-1-carboxylate
Quantity 1g
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Recommended Storage 2° to 8°C
Assay 98%
Molecular Formula C10H19NO2
MDL Number MFCD02093935
Beilstein 3604898
Boiling Point 130° to 140°C (15mmHg)
Flash Point 86°C (187°F)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

2° to 8°C