Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-isonipecotic acid ethyl ester, 97+%

$82.04 - $257.05

Chemical Identifiers

Chemical Name or Material 1-Boc-isonipecotic acid ethyl ester
CAS 142851-03-4
Molecular Formula C13H23NO4
Formula Weight 257.33
MDL Number MFCD01763998
Synonym Ethyl N-Boc-piperidine-4-carboxylate; Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2539306

  SDS
alfa aesar™
B2539306
5g Each for $82.04

AAB2539314

  SDS
alfa aesar™
B2539314
25g Each for $257.05
Description & Specifications

Specifications

Formula Weight 257.33
Chemical Name or Material 1-Boc-isonipecotic acid ethyl ester
Assay ≥97%
Molecular Formula C13H23NO4
MDL Number MFCD01763998
CAS 142851-03-4
Synonym Ethyl N-Boc-piperidine-4-carboxylate; Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester
Boiling Point 120° to 135°C (0.5mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C