Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-indoline, 98%

$27.57 - $72.18

Chemical Identifiers

Chemical Name or Material 1-Boc-indoline
CAS 143262-10-6
Molecular Formula C13H17NO2
Formula Weight 219.29
MDL Number MFCD01318399
Beilstein 5336249
Synonym 1-(tert-Butoxycarbonyl)indoline; tert-Butyl indoline-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1747706

  SDS
alfa aesar™
L1747706
5g Each for $72.18

AAL1747703

  SDS
alfa aesar™
L1747703
1g Each for $27.57
Description & Specifications

Specifications

Formula Weight 219.29
MDL Number MFCD01318399
Boiling Point 150°C (15mmHg)
Melting Point 45° to 49°C
Beilstein 5336249
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H17NO2
Synonym 1-(tert-Butoxycarbonyl)indoline; tert-Butyl indoline-1-carboxylate
Flash Point >110°C (230°F)
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Chemical Name or Material 1-Boc-indoline
Assay 98%
Health Hazard 1 Warning
CAS 143262-10-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C