missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-imidazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0050614

 View more versions of this product

Catalog No. AAL0050614


Description

Description

1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N?-carbonyldiimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.

Solubility
Soluble in hexane, acetone, and DMF.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Boc-imidazole
44°C to 48°C
MFCD00014497
607792
Soluble in hexane,acetone,and DMF.
CC(C)(C)OC(=O)N1C=CN=C1
168.196
168.2
49761-82-2
C8H12N2O2
25 g
n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p
MTBKGWHHOBJMHJ-UHFFFAOYSA-N
tert-butyl imidazole-1-carboxylate
521262
98%

Chemical Identifiers

49761-82-2
168.196
MTBKGWHHOBJMHJ-UHFFFAOYSA-N
521262
CC(C)(C)OC(=O)N1C=CN=C1
C8H12N2O2
MFCD00014497
n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p
tert-butyl imidazole-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 256-475-1

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only