missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-BOC-azetidine-3-methanol, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452090010

 View more versions of this product

Catalog No. AC452090010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-BOC-azetidine-3-methanol
Glass Bottle
1g
95% min. (GC)
1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, 1-boc-3-azetidinemethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-ylmethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, tert-butyl3-hydroxymethyl azetidine-1-carboxylate
CC(C)(C)OC(=O)N1CC(C1)CO
187.24
187.24
117°C
50°C to 53°C
142253-56-3
C9H17NO3
HXRDRJKAEYHOBB-UHFFFAOYSA-N
tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
10583745
96%

Chemical Identifiers

142253-56-3
187.24
1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, 1-boc-3-azetidinemethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-ylmethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, tert-butyl3-hydroxymethyl azetidine-1-carboxylate
tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
C9H17NO3
HXRDRJKAEYHOBB-UHFFFAOYSA-N
10583745
CC(C)(C)OC(=O)N1CC(C1)CO
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents