missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Boc-amino)cyclobutanecarboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6288806

 View more versions of this product

Catalog No. AAH6288806



Specifications

Specifications

1-(Boc-amino)cyclobutanecarboxylic acid
5g
C10H17NO4
7994003
ROVVUKFHORPDSM-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
1512646
Solid
129°C to 133°C
120728-10-1
MFCD02682623
n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
215.249
215.24
97%

Chemical Identifiers

120728-10-1
215.249
ROVVUKFHORPDSM-UHFFFAOYSA-N
1512646
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
C10H17NO4
MFCD02682623
n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents