Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-6-bromoindole, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-6-bromoindole
CAS 147621-26-9
Formula Weight 296.16
MDL Number MFCD09954826
Beilstein 9050272
Synonym tert-Butyl 6-bromoindole-1-carboxylate; 6-Bromoindole-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6353803

 
alfa aesar™
H6353803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6353806

 
alfa aesar™
H6353806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63538MD

 
alfa aesar™
H63538MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.16
Chemical Name or Material 1-Boc-6-bromoindole
Physical Form Solid
Assay 97%
MDL Number MFCD09954826
Quantity 1g
CAS 147621-26-9
Beilstein 9050272
Synonym tert-Butyl 6-bromoindole-1-carboxylate; 6-Bromoindole-1-carboxylic acid tert-butyl ester