Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-piperidone, 99%

$408.34 - $408.34

Chemical Identifiers

Chemical Name or Material 1-Boc-4-piperidone
CAS 79099-07-3
Molecular Formula C10H17NO3
Formula Weight 199.25
MDL Number MFCD00151800
Beilstein 3650236
Synonym 1-(tert-Butoxycarbonyl)-4-piperidone; tert-Butyl 4-oxo-1-piperidinecarboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1336118

  SDS
alfa aesar™
L1336118
50g Each for $408.34
Description & Specifications

Specifications

Formula Weight 199.25
Chemical Name or Material 1-Boc-4-piperidone
CAS 79099-07-3
Assay 99%
Molecular Formula C10H17NO3
MDL Number MFCD00151800
Beilstein 3650236
Synonym 1-(tert-Butoxycarbonyl)-4-piperidone; tert-Butyl 4-oxo-1-piperidinecarboxylate
Quantity 50g
Melting Point 72° to 76°C

Applications
1-Boc-4-piperidone is used as a pharmaceutical intermediate. It is involved in the preparation of 1-piperidin-4-yl-substituted butyro- and valerolactams.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and metals.