missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-piperidone, 99%

Manufacturer:  Alfa Aesar™ L1336118

 View more versions of this product

Catalog No. AAL1336118



Description

Description

Applications
1-Boc-4-piperidone is used as a pharmaceutical intermediate. It is involved in the preparation of 1-piperidin-4-yl-substituted butyro- and valerolactams.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and metals.
Specifications

Specifications

1-Boc-4-piperidone
50g
C10H17NO3
3650236
ROUYFJUVMYHXFJ-UHFFFAOYSA-N
tert-butyl 4-oxopiperidine-1-carboxylate
735900
99%
72°C to 76°C
79099-07-3
MFCD00151800
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
CC(C)(C)OC(=O)N1CCC(=O)CC1
199.25
199.25

Chemical Identifiers

79099-07-3
199.25
ROUYFJUVMYHXFJ-UHFFFAOYSA-N
735900
CC(C)(C)OC(=O)N1CCC(=O)CC1
C10H17NO3
MFCD00151800
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
tert-butyl 4-oxopiperidine-1-carboxylate
Documents