Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid, 96%

$100.51 - $344.80

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid
CAS 1210863-93-6
Molecular Formula C12H19NO6
Formula Weight 273.29
MDL Number MFCD07781134
Synonym 1-(tert-Butoxycarbonyl)-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid; Pyrrolidine-1,3,4-tricarboxylic acid 1-tert-butyl 3-methyl diester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5008603

 
alfa aesar™
H5008603
1g Each for $344.80

AAH50086MD

 
alfa aesar™
H50086MD
250mg Each for $100.51
Description & Specifications

Specifications

Formula Weight 273.29
Chemical Name or Material 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid
Assay 96%
Molecular Formula C12H19NO6
MDL Number MFCD07781134
CAS 1210863-93-6
Synonym 1-(tert-Butoxycarbonyl)-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid; Pyrrolidine-1,3,4-tricarboxylic acid 1-tert-butyl 3-methyl diester
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.