Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-cyanopiperidine, 96%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-cyanopiperidine
CAS 91419-52-2
Molecular Formula C11H18N2O2
Formula Weight 210.27
MDL Number MFCD01861223
UN Number UN3439
Synonym 1-Boc-piperidine-4-carbonitrile; tert-Butyl 4-cyanopiperidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2758003

  SDS
alfa aesar™
H2758003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2758006

  SDS
alfa aesar™
H2758006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27580MD

  SDS
alfa aesar™
H27580MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.27
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 1 Danger
Molecular Formula C11H18N2O2
CAS 91419-52-2
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01861223
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Melting Point 45° to 48°C
Assay 96%
Quantity 1g
Chemical Name or Material 1-Boc-4-cyanopiperidine
Synonym 1-Boc-piperidine-4-carbonitrile; tert-Butyl 4-cyanopiperidine-1-carboxylate
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.