missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(6-nitro-3-pyridyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5446506

 View more versions of this product

Catalog No. AAH5446506



Specifications

Specifications

1-Boc-4-(6-nitro-3-pyridyl)piperazine
571189-16-7
MFCD11849291
SUWKOEMQNOBJEQ-UHFFFAOYSA-N
tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
11243758
97%
5g
C14H20N4O4
1-boc-4-6-nitro-3-pyridyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CN=C(C=C2)[N+](=O)[O-]
308.338
308.33

Chemical Identifiers

571189-16-7
308.338
SUWKOEMQNOBJEQ-UHFFFAOYSA-N
11243758
CC(C)(C)OC(=O)N1CCN(CC1)C2=CN=C(C=C2)[N+](=O)[O-]
C14H20N4O4
MFCD11849291
1-boc-4-6-nitro-3-pyridyl piperazine
tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents