Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(5-iodo-2-pyridyl)piperazine, 95%

$79.24 - $286.28

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(5-iodo-2-pyridyl)piperazine
CAS 497915-42-1
Molecular Formula C14H20IN3O2
Formula Weight 389.24
MDL Number MFCD07781193
Synonym tert-Butyl 4-(5-iodopyridin-2-yl)piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5014403

  SDS
alfa aesar™
H5014403
1g Each for $286.28

AAH50144MD

  SDS
alfa aesar™
H50144MD
250mg Each for $79.24
Description & Specifications

Specifications

Formula Weight 389.24
Assay 95%
Molecular Formula C14H20IN3O2
Quantity 1g
Health Hazard 1 Warning
Synonym tert-Butyl 4-(5-iodopyridin-2-yl)piperazine-1-carboxylate
Chemical Name or Material 1-Boc-4-(5-iodo-2-pyridyl)piperazine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 497915-42-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD07781193
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.