missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole, 97%

Chemical Identifiers

CAS 1347815-06-8
Molecular Formula C15H13N5O2
Molecular Weight (g/mol) 295.302
MDL Number MFCD20265231
InChI Key VHYAIVBSALAEJF-UHFFFAOYSA-N
Synonym 1-boc-4,5-dicyano-2-3-pyridyl imidazole
PubChem CID 73995745
IUPAC Name tert-butyl 4,5-dicyano-2-pyridin-3-ylimidazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C(=C(N=C1C2=CN=CC=C2)C#N)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54303MD View Documents Alfa Aesar™
H54303MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5430303 View Documents Alfa Aesar™
H5430303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5430306 View Documents Alfa Aesar™
H5430306
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1347815-06-8
295.302
VHYAIVBSALAEJF-UHFFFAOYSA-N
73995745
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CN=CC=C2)C#N)C#N
C15H13N5O2
MFCD20265231
1-boc-4,5-dicyano-2-3-pyridyl imidazole
tert-butyl 4,5-dicyano-2-pyridin-3-ylimidazole-1-carboxylate

Specifications

250mg
C15H13N5O2
1-boc-4,5-dicyano-2-3-pyridyl imidazole
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CN=CC=C2)C#N)C#N
295.302
295.3
1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole
1347815-06-8
MFCD20265231
VHYAIVBSALAEJF-UHFFFAOYSA-N
tert-butyl 4,5-dicyano-2-pyridin-3-ylimidazole-1-carboxylate
73995745
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures