missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole, 97%

Manufacturer:  Alfa Aesar™ H5430303

 View more versions of this product

Catalog No. AAH5430303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole
1347815-06-8
MFCD20265231
VHYAIVBSALAEJF-UHFFFAOYSA-N
tert-butyl 4,5-dicyano-2-pyridin-3-ylimidazole-1-carboxylate
73995745
97%
1g
C15H13N5O2
1-boc-4,5-dicyano-2-3-pyridyl imidazole, tert-butyl 4,5-dicyano-2-pyridin-3-yl imidazole-1-carboxylate
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CN=CC=C2)C#N)C#N
295.302
295.3

Chemical Identifiers

1347815-06-8
295.302
VHYAIVBSALAEJF-UHFFFAOYSA-N
73995745
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CN=CC=C2)C#N)C#N
C15H13N5O2
MFCD20265231
1-boc-4,5-dicyano-2-3-pyridyl imidazole, tert-butyl 4,5-dicyano-2-pyridin-3-yl imidazole-1-carboxylate
tert-butyl 4,5-dicyano-2-pyridin-3-ylimidazole-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents