missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6361503

 View more versions of this product

Catalog No. AAH6361503



Specifications

Specifications

1g
C15H21N3O4
1-4-boc-piperazin-1-yl-4-nitrobenzene, 1-boc-4-4-nitrophenyl piperazine, 1-n-boc-4-4-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-nitrophenyl-, 1,1-dimethylethyl ester, 1-t-butoxycarbonyl-4-4-nitrophenyl piperazine, 1-t-butoxycarbonyl4-4-nitrophenyl piperazine, 4-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester, acmc-1c6wn, tert-butyl 4-4-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-4-nitrophenyl piperazinecarboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
307.35
307.34
97%
182618-86-6
MFCD02187326
MGYCIJUTYLUYJM-UHFFFAOYSA-N
tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
3380696
Solid

Chemical Identifiers

182618-86-6
307.35
MGYCIJUTYLUYJM-UHFFFAOYSA-N
3380696
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
C15H21N3O4
MFCD02187326
1-4-boc-piperazin-1-yl-4-nitrobenzene, 1-boc-4-4-nitrophenyl piperazine, 1-n-boc-4-4-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-nitrophenyl-, 1,1-dimethylethyl ester, 1-t-butoxycarbonyl-4-4-nitrophenyl piperazine, 1-t-butoxycarbonyl4-4-nitrophenyl piperazine, 4-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester, acmc-1c6wn, tert-butyl 4-4-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-4-nitrophenyl piperazinecarboxylate
tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents