missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-formylphenyl)piperazine, 97%

$81.64 - $1,247.19

Chemical Identifiers

CAS 197638-83-8
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.363
MDL Number MFCD05864663
InChI Key KHORERZDMJTBMR-UHFFFAOYSA-N
Synonym 1-boc-4-4-formylphenyl piperazine
PubChem CID 2795509
IUPAC Name tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5541303 View Documents Alfa Aesar™
H5541303
1g Each for $81.64
 
AAH5541306 View Documents Alfa Aesar™
H5541306
5g Each for $311.80
 
AAH5541314 View Documents Alfa Aesar™
H5541314
25g Each for $1,247.19
 
Specifications

Chemical Identifiers

197638-83-8
290.363
KHORERZDMJTBMR-UHFFFAOYSA-N
2795509
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C=O
C16H22N2O3
MFCD05864663
1-boc-4-4-formylphenyl piperazine
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate

Specifications

Air Sensitive
1g
C16H22N2O3
UN2811
KHORERZDMJTBMR-UHFFFAOYSA-N
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
2795509
97%
118°C to 121°C
197638-83-8
MFCD05864663
1-boc-4-4-formylphenyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C=O
290.363
290.36
1-Boc-4-(4-formylphenyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen