missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-formylphenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5541314

 View more versions of this product

Catalog No. AAH5541314



Specifications

Specifications

1-Boc-4-(4-formylphenyl)piperazine
118°C to 121°C
197638-83-8
MFCD05864663
1-boc-4-4-formylphenyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C=O
290.363
290.36
Air Sensitive
25g
C16H22N2O3
UN2811
KHORERZDMJTBMR-UHFFFAOYSA-N
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
2795509
97%

Chemical Identifiers

197638-83-8
290.363
KHORERZDMJTBMR-UHFFFAOYSA-N
2795509
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C=O
C16H22N2O3
MFCD05864663
1-boc-4-4-formylphenyl piperazine
tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents