missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 874814-94-5
Molecular Formula C15H20FN3O4
Molecular Weight (g/mol) 325.34
MDL Number MFCD06408840
InChI Key VQQDLDPXBVJQDP-UHFFFAOYSA-N
Synonym tert-butyl 4-4-fluoro-2-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-4-fluoro-2-nitrophenyl-piperazine-1-carboxylate, 1-boc-4-4-fluoro-2-nitrophenyl piperazine
PubChem CID 51301635
IUPAC Name tert-butyl 4-(4-fluoro-2-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63438MD View Documents Alfa Aesar™
H63438MD
250mg Each for $35.93
 
AAH6343803 View Documents Alfa Aesar™
H6343803
1g Each for $106.26
 
AAH6343806 View Documents Alfa Aesar™
H6343806
5g Each for $422.54
 
Specifications

Chemical Identifiers

874814-94-5
325.34
VQQDLDPXBVJQDP-UHFFFAOYSA-N
51301635
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
C15H20FN3O4
MFCD06408840
tert-butyl 4-4-fluoro-2-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-4-fluoro-2-nitrophenyl-piperazine-1-carboxylate, 1-boc-4-4-fluoro-2-nitrophenyl piperazine
tert-butyl 4-(4-fluoro-2-nitrophenyl)piperazine-1-carboxylate

Specifications

250mg
C15H20FN3O4
18325431
VQQDLDPXBVJQDP-UHFFFAOYSA-N
tert-butyl 4-(4-fluoro-2-nitrophenyl)piperazine-1-carboxylate
51301635
Solid
1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine
874814-94-5
MFCD06408840
tert-butyl 4-4-fluoro-2-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-4-fluoro-2-nitrophenyl-piperazine-1-carboxylate, 1-boc-4-4-fluoro-2-nitrophenyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
325.34
325.34
97%