missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6343806

 View more versions of this product

Catalog No. AAH6343806


 

Specifications

Specifications

1-Boc-4-(4-Fluoro-2-nitrophenyl)piperazine
97%
MFCD06408840
18325431
VQQDLDPXBVJQDP-UHFFFAOYSA-N
tert-butyl 4-(4-fluoro-2-nitrophenyl)piperazine-1-carboxylate
51301635
Solid
874814-94-5
C15H20FN3O4
5g
tert-Butyl 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylate; 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
325.34
325.34

Chemical Identifiers

874814-94-5
325.34
VQQDLDPXBVJQDP-UHFFFAOYSA-N
51301635
C15H20FN3O4
MFCD06408840
tert-Butyl 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylate; 4-(4-Fluoro-2-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
Documents