missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-chloro-2-nitrophenyl)piperazine, 97%

$35.93 - $465.72

Chemical Identifiers

CAS 874814-97-8
Molecular Formula C15H20ClN3O4
Molecular Weight (g/mol) 341.792
InChI Key OJIDLANQYRRUCV-UHFFFAOYSA-N
Synonym tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
PubChem CID 69042278
IUPAC Name tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63592MD View Documents Alfa Aesar™
H63592MD
250mg Each for $35.93
 
AAH6359203 View Documents Alfa Aesar™
H6359203
1g Each for $106.26
 
AAH6359206 View Documents Alfa Aesar™
H6359206
5g Each for $465.72
 
Specifications

Chemical Identifiers

874814-97-8
341.792
tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
C15H20ClN3O4
OJIDLANQYRRUCV-UHFFFAOYSA-N
69042278
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

250mg
C15H20ClN3O4
tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
341.792
341.79
97%
874814-97-8
18477062
OJIDLANQYRRUCV-UHFFFAOYSA-N
tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
69042278
Solid
1-Boc-4-(4-chloro-2-nitrophenyl)piperazine