missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(4-chloro-2-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6359206

 View more versions of this product

Catalog No. AAH6359206



Specifications

Specifications

1-Boc-4-(4-chloro-2-nitrophenyl)piperazine
874814-97-8
18477062
OJIDLANQYRRUCV-UHFFFAOYSA-N
tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
69042278
Solid
5g
C15H20ClN3O4
tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
341.792
341.79
97%

Chemical Identifiers

874814-97-8
341.792
tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
C15H20ClN3O4
OJIDLANQYRRUCV-UHFFFAOYSA-N
69042278
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
Documents