Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(3-iodo-2-pyridyl)piperazine, 95%

$410.36 - $410.36

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(3-iodo-2-pyridyl)piperazine
CAS 902837-43-8
Molecular Formula C14H20IN3O2
Formula Weight 389.24g/mol
MDL Number MFCD07781155
Synonym tert-Butyl 4-(3-iodopyridin-2-yl)piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5005706

 
alfa aesar™
H50057-06
Each for $410.36
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 1-Boc-4-(3-iodo-2-pyridyl)piperazine
CAS 902837-43-8
Assay 95%
Molecular Formula C14H20IN3O2
Formula Weight 389.24g/mol
MDL Number MFCD07781155
Synonym tert-Butyl 4-(3-iodopyridin-2-yl)piperazine-1-carboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.