Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
CAS 261925-94-4
Formula Weight 334.42
MDL Number MFCD07369778
UN Number UN2811
Synonym 1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5559806

  SDS
alfa aesar™
H5559806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5559814

  SDS
alfa aesar™
H5559814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 62° to 66°C
Formula Weight 334.42
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Physical Form Powder
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
MDL Number MFCD07369778
Synonym 1-tert-Butoxycarbonyl-4-[3-(ethoxycarbonyl)phenyl]piperazine
Color Yellow to orange
Assay 97%
Chemical Name or Material 1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
CAS 261925-94-4