missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-nitrophenyl)piperazine, 97%

$28.84 - $371.65

Chemical Identifiers

CAS 170017-73-9
Molecular Formula C15H21N3O4
Molecular Weight (g/mol) 307.35
MDL Number MFCD08275017
InChI Key GTGIZVUOLBRDAO-UHFFFAOYSA-N
Synonym 1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
PubChem CID 21935210
IUPAC Name tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63876MD View Documents Alfa Aesar™
H63876MD
250mg Each for $28.84
 
AAH6387603 View Documents Alfa Aesar™
H6387603
1g Each for $85.14
 
AAH6387606 View Documents Alfa Aesar™
H6387606
5g Each for $371.65
 
Specifications

Chemical Identifiers

170017-73-9
307.35
GTGIZVUOLBRDAO-UHFFFAOYSA-N
21935210
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
C15H21N3O4
MFCD08275017
1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate

Specifications

250mg
C15H21N3O4
1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
307.35
307.34
97%
170017-73-9
MFCD08275017
GTGIZVUOLBRDAO-UHFFFAOYSA-N
tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate
21935210
Solid
1-Boc-4-(2-nitrophenyl)piperazine