missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6387606

 View more versions of this product

Catalog No. AAH6387606



Specifications

Specifications

1-Boc-4-(2-nitrophenyl)piperazine
170017-73-9
MFCD08275017
GTGIZVUOLBRDAO-UHFFFAOYSA-N
tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate
21935210
Solid
5g
C15H21N3O4
1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
307.35
307.34
97%

Chemical Identifiers

170017-73-9
307.35
GTGIZVUOLBRDAO-UHFFFAOYSA-N
21935210
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
C15H21N3O4
MFCD08275017
1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate
Documents