missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-methoxycarbonylphenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5530203

 View more versions of this product

Catalog No. AAH5530203



Specifications

Specifications

1-Boc-4-(2-methoxycarbonylphenyl)piperazine
1.532
1g
C17H24N2O4
UN2810
JFZGCXORLWXLGH-UHFFFAOYSA-N
tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
16217924
Liquid
>110°C (230°F)
Yellow to Orange
870703-74-5
MFCD07369760
1-boc-4-2-methoxycarbonyl phenyl-piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C(=O)OC
320.389
320.39
97%

Chemical Identifiers

870703-74-5
320.389
JFZGCXORLWXLGH-UHFFFAOYSA-N
16217924
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C(=O)OC
C17H24N2O4
MFCD07369760
1-boc-4-2-methoxycarbonyl phenyl-piperazine
tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents