Learn More
1-Boc-4-(2-formylphenyl)piperazine, 97%, Thermo Scientific™
$84.88 - $1336.27
Chemical Identifiers
CAS | 174855-57-3 |
---|---|
Molecular Formula | C16H22N2O3 |
Molecular Weight (g/mol) | 290.363 |
MDL Number | MFCD05864664 |
InChI Key | FGJACYJASSSXNJ-UHFFFAOYSA-N |
Synonym | 1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine |
PubChem CID | 6490990 |
IUPAC Name | tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5596603
|
Thermo Scientific Chemicals
H5596603 |
1 g |
Each for $84.88
|
|
|||||
AAH5596614
|
Thermo Scientific Chemicals
H5596614 |
25 g |
Each for $1,336.27
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
174855-57-3 | |
290.363 | |
FGJACYJASSSXNJ-UHFFFAOYSA-N | |
6490990 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O |
C16H22N2O3 | |
MFCD05864664 | |
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine | |
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate |
Specifications
174855-57-3 | |
C16H22N2O3 | |
1 g | |
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O | |
290.363 | |
290.36 | |
1-Boc-4-(2-formylphenyl)piperazine |
86°C to 90°C | |
MFCD05864664 | |
UN2811 | |
FGJACYJASSSXNJ-UHFFFAOYSA-N | |
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate | |
6490990 | |
97% |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures