missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-4-(2-formylphenyl)piperazine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5596614

 View more versions of this product

Catalog No. AAH5596614



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Boc-4-(2-formylphenyl)piperazine
86°C to 90°C
MFCD05864664
UN2811
FGJACYJASSSXNJ-UHFFFAOYSA-N
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
6490990
97%
174855-57-3
C16H22N2O3
25 g
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
290.363
290.36

Chemical Identifiers

174855-57-3
290.363
FGJACYJASSSXNJ-UHFFFAOYSA-N
6490990
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
C16H22N2O3
MFCD05864664
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions