missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-formylphenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5596606

 View more versions of this product

Catalog No. AAH5596606



Description

Description

Specifications

Specifications

1-Boc-4-(2-formylphenyl)piperazine
5g
C16H22N2O3
UN2811
FGJACYJASSSXNJ-UHFFFAOYSA-N
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
6490990
97%
86°C to 90°C
174855-57-3
MFCD05864664
1-boc-4-2-formyl-phenyl-piperazine, 1-boc-4-2-formylphenyl piperazine, 1-n-boc-4-2-formylphenyl piperazine, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, tert-butyl 4-2-formylphenyl piperazinecarboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
290.363
290.36

Chemical Identifiers

174855-57-3
290.363
FGJACYJASSSXNJ-UHFFFAOYSA-N
6490990
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
C16H22N2O3
MFCD05864664
1-boc-4-2-formyl-phenyl-piperazine, 1-boc-4-2-formylphenyl piperazine, 1-n-boc-4-2-formylphenyl piperazine, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, tert-butyl 4-2-formylphenyl piperazinecarboxylate
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents