missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 1233958-36-5
Molecular Formula C15H20FN3O4
Molecular Weight (g/mol) 325.34
MDL Number MFCD14155993
InChI Key GWHUVEIGDXIWOY-UHFFFAOYSA-N
Synonym tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine, tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate
PubChem CID 53255503
IUPAC Name tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63668MD View Documents Alfa Aesar™
H63668MD
250mg Each for $35.93
 
AAH6366803 View Documents Alfa Aesar™
H6366803
1g Each for $106.26
 
AAH6366806 View Documents Alfa Aesar™
H6366806
5g Each for $422.54
 
Specifications

Chemical Identifiers

1233958-36-5
325.34
GWHUVEIGDXIWOY-UHFFFAOYSA-N
53255503
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
C15H20FN3O4
MFCD14155993
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine, tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate

Specifications

250mg
C15H20FN3O4
19933459
GWHUVEIGDXIWOY-UHFFFAOYSA-N
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
53255503
Solid
1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine
1233958-36-5
MFCD14155993
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine, tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
325.34
325.34
97%