missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6366806

 View more versions of this product

Catalog No. AAH6366806


 

Specifications

Specifications

1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine
97%
MFCD14155993
19933459
GWHUVEIGDXIWOY-UHFFFAOYSA-N
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
53255503
Solid
1233958-36-5
C15H20FN3O4
5g
tert-Butyl 4-(2-Fluoro-6-nitrophenyl)piperazine-1-carboxylate; 4-(2-Fluoro-6-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
325.34
325.34

Chemical Identifiers

1233958-36-5
325.34
GWHUVEIGDXIWOY-UHFFFAOYSA-N
53255503
C15H20FN3O4
MFCD14155993
tert-Butyl 4-(2-Fluoro-6-nitrophenyl)piperazine-1-carboxylate; 4-(2-Fluoro-6-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
Documents