Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine
CAS 154590-34-8
Formula Weight 325.34
MDL Number MFCD08443959
Synonym tert-Butyl 4-(2-Fluoro-4-nitrophenyl)piperazine-1-carboxylate; 4-(2-Fluoro-4-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6334003

 
alfa aesar™
H6334003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6334006

 
alfa aesar™
H6334006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63340MD

 
alfa aesar™
H63340MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 325.34
Chemical Name or Material 1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine
Physical Form Solid
Assay 97%
MDL Number MFCD08443959
Quantity 1g
CAS 154590-34-8
Synonym tert-Butyl 4-(2-Fluoro-4-nitrophenyl)piperazine-1-carboxylate; 4-(2-Fluoro-4-nitrophenyl)piperazine-1-carboxylic acid tert-butyl ester