missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H63340MD

 View more versions of this product

Catalog No. AAH63340MD



Specifications

Specifications

1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine
154590-34-8
MFCD08443959
YOLVWCABLVHASL-UHFFFAOYSA-N
tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate
16203508
Solid
250mg
C15H20FN3O4
tert-butyl 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-2-fluoro-4-nitrophenyl tetrahydro-1 2h-pyrazinecarboxylate, butylfluoronitrophenyltetrahydropyrazinecarboxylate, 1-boc-4-2-fluoro-4-nitrophenyl piperazine, 1-tert-butoxycarbonyl-4-2-fluoro-4-nitrophenyl piperazine, 3-fluoro-4-4-tert-butoxycarbonyl piperazin-1-yl nitrobenzene, tert-butyl4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, 4-2-fluoro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitro-phenyl piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
325.34
325.34
97%

Chemical Identifiers

154590-34-8
325.34
YOLVWCABLVHASL-UHFFFAOYSA-N
16203508
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
C15H20FN3O4
MFCD08443959
tert-butyl 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-2-fluoro-4-nitrophenyl tetrahydro-1 2h-pyrazinecarboxylate, butylfluoronitrophenyltetrahydropyrazinecarboxylate, 1-boc-4-2-fluoro-4-nitrophenyl piperazine, 1-tert-butoxycarbonyl-4-2-fluoro-4-nitrophenyl piperazine, 3-fluoro-4-4-tert-butoxycarbonyl piperazin-1-yl nitrobenzene, tert-butyl4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, 4-2-fluoro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitro-phenyl piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester
tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate
Documents