Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-chloro-6-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(2-chloro-6-nitrophenyl)piperazine
CAS 400803-05-6
Formula Weight 341.79
Beilstein 14092196
Synonym tert-Butyl 4-(2-chloro-6-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-6-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6312703

 
alfa aesar™
H6312703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6312706

 
alfa aesar™
H6312706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63127MD

 
alfa aesar™
H63127MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.79
Synonym tert-Butyl 4-(2-chloro-6-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-6-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
Quantity 1g
Chemical Name or Material 1-Boc-4-(2-chloro-6-nitrophenyl)piperazine
Physical Form Solid
CAS 400803-05-6
Assay 97%
Beilstein 14092196